3-Methoxy-1-propanol, 98%
3-Methoxy-1-propanol, 98%
3-Methoxy-1-propanol, 98%
Thermo Scientific Chemicals

3-Methoxy-1-propanol, 98%

CAS: 1589-49-7 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 137150500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1589-49-7
IUPAC Name3-methoxypropan-1-ol
Molecular FormulaC4H10O2
InChI KeyJDFDHBSESGTDAL-UHFFFAOYSA-N
SMILESCOCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
GC>=97.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only