Lidocaine, 97.5%
Lidocaine, 97.5%
Lidocaine, 97.5%
Thermo Scientific Chemicals

Lidocaine, 97.5%

CAS: 137-58-6 | C14H22N2O | 234.343 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 137251000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS137-58-6
IUPAC Name2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
Molecular FormulaC14H22N2O
InChI KeyNNJVILVZKWQKPM-UHFFFAOYSA-N
SMILESCCN(CC)CC(=O)NC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Melting point66°C to 69°C
Titration with HClO4>=96.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only