Product Image
Invitrogen™
BLOCK-iT™ Transfection Optimization Kit
The BLOCK-iT™ Transfection Optimization Kit is designed to optimize RNAi transfection using controls for transfection and viability. Each kit containsRead more
Have Questions?
Catalog NumberQuantity
137500471 Kit
Catalog number 13750047
Price (USD)
792.00
Each
Quantity:
1 Kit
Price (USD)
792.00
Each
The BLOCK-iT™ Transfection Optimization Kit is designed to optimize RNAi transfection using controls for transfection and viability. Each kit contains a BLOCK-iT™ Fluorescent Oligo, which provides:
• A strong, easily detectable FITC signal indicating transfection efficiency
• Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA provides
• Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules
This makes BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. In addition, the kit contains a Positive Control Stealth RNAi™ siRNA molecule directed at human p53 and a Scrambled Stealth RNAi™ siRNA molecule as a negative control. To assay cell viability, Dead Cell Dye is included. This dye colors dead cells as red, thereby allowing easy identification of non-viable cells in a tissue culture plate. Using these reagents together allows you to both optimize transfection conditions and have a functional positive control in every RNAi experiment you perform.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Control TypeCell Viability Control, Negative Control, Transfection Control, Positive Control
For Use With (Application)Transfection
Product LineBLOCK-iT™, Stealth RNAi™ siRNA
Product TypeTransfection Kit
Quantity1 Kit
RNAi TypesiRNA
FormatKit
Sample TypeRNA
VectorpLATE
Unit SizeEach
Citations & References (1)
Citations & References
Abstract
RNAi specificity: how big of an issue is it?
Authors:Samarsky DA, Welch PJ,
Journal:Pharmacogenomics
PubMed ID:15723598
RNAi techniques hold great promise for science, from basic research all the way to human therapeutics, and our understanding of the underlying mechanisms have allowed us to design better and more effective compounds. Fortunately, much has also been learned about nonspecific effects, such as off-target and stress responses. These observations and discoveries have led us to improved specificity ... More