D(-)-Tartaric acid, 99%
D(-)-Tartaric acid, 99%
D(-)-Tartaric acid, 99%
Thermo Scientific Chemicals

D(-)-Tartaric acid, 99%

CAS: 147-71-7 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 137860010
Price (EUR)
966,00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
966,00
Each
Chemical Identifiers
CAS147-71-7
IUPAC Name2,3-dihydroxybutanedioic acid
Molecular FormulaC4H6O6
InChI KeyFEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILESOC(C(O)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point167°C to 171°C
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only