Terephthaldicarboxaldehyde, 98%
Terephthaldicarboxaldehyde, 98%
Terephthaldicarboxaldehyde, 98%
Acros Organics

Terephthaldicarboxaldehyde, 98%

CAS: 623-27-8 | C8H6O2 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 137891000
also known as 13789
HSN 29122990
Price (INR)
10,069.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
10,069.00
Each
Chemical Identifiers
CAS623-27-8
IUPAC Namebenzene-1,4-dicarbaldehyde
Molecular FormulaC8H6O2
InChI KeyKUCOHFSKRZZVRO-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point114°C to 116°C
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only