p-Terphenyl, 99+%, pure
p-Terphenyl, 99+%, pure
p-Terphenyl, 99+%, pure
Thermo Scientific Chemicals

p-Terphenyl, 99+%, pure

CAS: 92-94-4 | C18H14 | 230.31 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 137920250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS92-94-4
IUPAC Name4-phenyl-1,1'-biphenyl
Molecular FormulaC18H14
InChI KeyXJKSTNDFUHDPQJ-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to light beige
Infrared spectrumConforms
Melting point211°C to 216°C
GC>=99.0 %
Appearance (Form)flakes
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only