2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%
2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%
2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%
Thermo Scientific Chemicals

2,4,5,6-Tetraaminopyrimidine sulfate hydrate, 98%

CAS: 5392-28-9 | C4H10N6O4S | 238.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 137930050
Price (USD)
14.50
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
14.50
Each
Chemical Identifiers
CAS5392-28-9
IUPAC Namepyrimidine-2,4,5,6-tetramine; sulfuric acid
Molecular FormulaC4H10N6O4S
InChI KeyMQEFDQWUCTUJCP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 % (On dry substance)
Loss on drying=<9 % (1 g, 150°C, 3 hrs)
Appearance (Color)Ochre to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only