alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%
alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%
alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%
Thermo Scientific Chemicals

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%

CAS: 13209-15-9 | C8H6Br4 | 421.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 137970250
Price (EUR)
30,65
Online exclusive
33,70
Save 3,05
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
30,65
Online exclusive
33,70
Save 3,05
Each
Chemical Identifiers
CAS13209-15-9
IUPAC Name1,2-bis(dibromomethyl)benzene
Molecular FormulaC8H6Br4
InChI KeyLNAOKZKISWEZNY-UHFFFAOYSA-N
SMILESBrC(Br)C1=CC=CC=C1C(Br)Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to light grey
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point114°C to 118°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only