Tetrachloroethylene, 99%, extra pure
Tetrachloroethylene, 99%, extra pure
Tetrachloroethylene, 99%, extra pure
Thermo Scientific Chemicals

Tetrachloroethylene, 99%, extra pure

Tetrachloroethylene, 99%, C2Cl4, CAS Number-127-18-4, ankilostin, carbon dichloride, ethene, tetrachloro, ethylene tetrachloride, perc, perchlorethylene, perchloroethylene, tetrachlorethylene, tetrachloroethene, tetrachloroethylene, 1L, 0.003% max. (NaOH), 98.5% min. (GC), 01, 187 | CAS: 127-18-4 | C2Cl4 | 165.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 138010100
Price (USD)
583.65
Special offer
Online exclusive
Ends: 28-Dec-2026
687.00
Save 103.35
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
583.65
Special offer
Online exclusive
Ends: 28-Dec-2026
687.00
Save 103.35
Each
Chemical Identifiers
CAS127-18-4
IUPAC Nametetrachloroethene
Molecular FormulaC2Cl4
InChI KeyCYTYCFOTNPOANT-UHFFFAOYSA-N
SMILESClC(Cl)=C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.01 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
GC>=98.5 %
Residue after evaporation=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only