1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
1-Tetradecylamine, 98%
Thermo Scientific Chemicals

1-Tetradecylamine, 98%

CAS: 2016-42-4 | C14H31N | 213.41 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
產品號碼 138100050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2016-42-4
IUPAC Namehydrogen tetradecan-1-amine chloride
Molecular FormulaC14H32ClN
InChI KeyKKWUACQXLWHLCX-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCCCCCCCCCCCCCN
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to pink
Titration with HClO4>=97.5 %
Appearance (Form)Crystals or crystalline solid
Infrared spectrumConforms
Melting point37°C to 42°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only