Specifications
Diameter (Metric) Outer38mm
For Use With (Application)For diluting alkalis, drawn from one piece of heavy pure sheet nickel and trimmed to final form
Height (Metric)32mm
Max. Temperature (Metric)800°C
Capacity (Metric)20 mL
MaterialNickel
Product TypeCrucible
Unit SizeEach