Tetramethylammonium hydroxide, 25 wt% in methanol
Tetramethylammonium hydroxide, 25 wt% in methanol
Tetramethylammonium hydroxide, 25 wt% in methanol
Thermo Scientific Chemicals

Tetramethylammonium hydroxide, 25 wt% in methanol

CAS: 75-59-2 | C4H13NO | 91.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 138351000
Price (USD)
68.50
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
68.50
Each
Chemical Identifiers
CAS75-59-2
IUPAC Nametetramethylazanium hydroxide
Molecular FormulaC4H13NO
InChI KeyWGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl24.5 to 26.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only