Tetramethylene sulfoxide, 97%
Tetramethylene sulfoxide, 97%
Tetramethylene sulfoxide, 97%
Thermo Scientific Chemicals

Tetramethylene sulfoxide, 97%

CAS: 1600-44-8 | C4H8OS | 104.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 138441000
Price (EUR)
215,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
215,00
Each
Chemical Identifiers
CAS1600-44-8
IUPAC Name1λ⁴-thiolan-1-one
Molecular FormulaC4H8OS
InChI KeyISXOBTBCNRIIQO-UHFFFAOYSA-N
SMILESO=S1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5190 to 1.5230 (20°C, 589 nm)
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only