Thiocarbanilide, 98%
Thiocarbanilide, 98%
Thiocarbanilide, 98%
Thermo Scientific Chemicals

Thiocarbanilide, 98%

CAS: 102-08-9 | C13H12N2S | 228.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 138700025
Price (EUR)
205,00
Each
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (EUR)
205,00
Each
Chemical Identifiers
CAS102-08-9
IUPAC Name1,3-diphenylthiourea
Molecular FormulaC13H12N2S
InChI KeyFCSHMCFRCYZTRQ-UHFFFAOYSA-N
SMILESS=C(NC1=CC=CC=C1)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point148°C to 154°C
Appearance (Color)White to cream to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only