p-Tolualdehyde, 97%
p-Tolualdehyde, 97%
p-Tolualdehyde, 97%
Thermo Scientific Chemicals

p-Tolualdehyde, 97%

CAS: 104-87-0 | C8H8O | 120.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
產品號碼 139000025
價格 (TWD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS104-87-0
IUPAC Name4-methylbenzaldehyde
Molecular FormulaC8H8O
InChI KeyFXLOVSHXALFLKQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=O)C=C1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Liquid
Refractive index1.5440 to 1.5480 (20°C, 589 nm)
GC>=96.0 %
Specific gravity(20°C) 1.010 to 1.025
Infrared spectrumConforms
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only