p-Toluenesulfinic acid, sodium salt hydrate, 98+%
p-Toluenesulfinic acid, sodium salt hydrate, 98+%
p-Toluenesulfinic acid, sodium salt hydrate, 98+%
Thermo Scientific Chemicals

p-Toluenesulfinic acid, sodium salt hydrate, 98+%

CAS: 207801-20-5 | C7H7NaO2S | 178.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 139011000
Price (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS207801-20-5
IUPAC Namesodium 4-methylbenzene-1-sulfinate
Molecular FormulaC7H7NaO2S
InChI KeyKFZUDNZQQCWGKF-UHFFFAOYSA-M
SMILES[Na+].CC1=CC=C(C=C1)S([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Titration with HClO4>=98.0 % (On dry substance)
Water=<5 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only