p-Toluenesulfonic acid monohydrate, 99%, extra pure
p-Toluenesulfonic acid monohydrate, 99%, extra pure
p-Toluenesulfonic acid monohydrate, 99%, extra pure
Acros Organics

p-Toluenesulfonic acid monohydrate, 99%, extra pure

CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 139021000
HSN 29041010
Price (INR)
5,698.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
5,698.00
Each
Chemical Identifiers
CAS6192-52-5
IUPAC Name4-methylbenzene-1-sulfonic acid hydrate
Molecular FormulaC7H10O4S
InChI KeyKJIFKLIQANRMOU-UHFFFAOYSA-N
SMILESO.CC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with CHA>=98.5 % (On dry substance)
Water10.0 to 13.0 % (KF)
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Titration with CHA=<0.5 % Sulfuric acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only