p-Toluic acid, 98%
p-Toluic acid, 98%
p-Toluic acid, 98%
Thermo Scientific Chemicals

p-Toluic acid, 98%

CAS: 99-94-5 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 139062500
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
56.30
Save 8.65
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
56.30
Save 8.65
Each
Chemical Identifiers
CAS99-94-5
IUPAC Name4-methylbenzoic acid
Molecular FormulaC8H8O2
InChI KeyLPNBBFKOUUSUDB-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point179°C to 182°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only