p-Tolylacetic acid, 99%
p-Tolylacetic acid, 99%
p-Tolylacetic acid, 99%
Thermo Scientific Chemicals

p-Tolylacetic acid, 99%

CAS: 622-47-9 | C9H10O2 | 150.18 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 139121000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS622-47-9
IUPAC Name2-(4-methylphenyl)acetate
Molecular FormulaC9H9O2
InChI KeyGXXXUZIRGXYDFP-UHFFFAOYSA-M
SMILESCC1=CC=C(CC([O-])=O)C=C1
さらに表示
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point90°C to 93°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only