Tributylamine, 99%
Tributylamine, 99%
Tributylamine, 99%
Thermo Scientific Chemicals

Tributylamine, 99%

CAS: 102-82-9 | C12H27N | 185.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 139320100
Price (USD)
317.65
Special offer
Online exclusive
Ends: 28-Dec-2026
374.00
Save 56.35
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
317.65
Special offer
Online exclusive
Ends: 28-Dec-2026
374.00
Save 56.35
Each
Chemical Identifiers
CAS102-82-9
IUPAC Nametributylamine
Molecular FormulaC12H27N
InChI KeyIMFACGCPASFAPR-UHFFFAOYSA-N
SMILESCCCCN(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Color scale=<20 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4273 to 1.4293 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only