Tri-n-butylphosphine, 95%
Tri-n-butylphosphine, 95%
Tri-n-butylphosphine, 95%
Thermo Scientific Chemicals

Tri-n-butylphosphine, 95%

CAS: 998-40-3 | C12H27P | 202.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 139345000
Price (USD)
282.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
282.00
Each
Chemical Identifiers
CAS998-40-3
IUPAC Nametributylphosphane
Molecular FormulaC12H27P
InChI KeyTUQOTMZNTHZOKS-UHFFFAOYSA-N
SMILESCCCCP(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
GC>=94 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only