1,1,2-Trichloroethane, 98%
1,1,2-Trichloroethane, 98%
1,1,2-Trichloroethane, 98%
Thermo Scientific Chemicals

1,1,2-Trichloroethane, 98%

CAS: 79-00-5 | C2H3Cl3 | 133.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 139432500
Price (EUR)
86,10
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
86,10
Each
Chemical Identifiers
CAS79-00-5
IUPAC Name1,1,2-trichloroethane
Molecular FormulaC2H3Cl3
InChI KeyUBOXGVDOUJQMTN-UHFFFAOYSA-N
SMILESClCC(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4700 to 1.4720 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only