2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
Thermo Scientific Chemicals

2,4,6-Trichlorophenol, 98%

CAS: 88-06-2 | C6H3Cl3O | 197.45 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
產品號碼 139485000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS88-06-2
IUPAC Name2,4,6-trichlorophenol
Molecular FormulaC6H3Cl3O
InChI KeyLINPIYWFGCPVIE-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=C(Cl)C=C1Cl
檢視更多
規格Specification Sheet規格表
Appearance (Form)Crystalline powder or mass and/or chunks
GC>=97.5 %
Infrared spectrumConforms
Melting point64°C to 68°C
Appearance (Color)Light beige to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only