2,4,6-Trichloropyrimidine, 99%
2,4,6-Trichloropyrimidine, 99%
2,4,6-Trichloropyrimidine, 99%
Thermo Scientific Chemicals

2,4,6-Trichloropyrimidine, 99%

CAS: 3764-01-0 | C4HCl3N2 | 183.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 139491000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3764-01-0
IUPAC Name2,4,6-trichloropyrimidine
Molecular FormulaC4HCl3N2
InChI KeyDPVIABCMTHHTGB-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=NC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Refractive index1.5690 to 1.5710 (20°C, 589 nm)
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only