Triethanolamine, 97%, Thermo Scientific Chemicals
Triethanolamine, 97%, Thermo Scientific Chemicals
Triethanolamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethanolamine, 97%, Thermo Scientific Chemicals

数量:
100 mL
1 L
2.5 L
25 L
製品番号(カタログ番号) 139561000
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS102-71-6
IUPAC Name2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Molecular FormulaC6H15NO3
InChI KeyGSEJCLTVZPLZKY-UHFFFAOYSA-N
SMILESOCCN(CCO)CCO
さらに表示
Color scale=<50 APHA
Refractive index1.4830 to 1.4860 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only