Triethyl methanetricarboxylate, 98%
Triethyl methanetricarboxylate, 98%
Triethyl methanetricarboxylate, 98%
Thermo Scientific Chemicals

Triethyl methanetricarboxylate, 98%

CAS: 6279-86-3 | C10H16O6 | 232.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 139630250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6279-86-3
IUPAC Nametriethyl methanetricarboxylate
Molecular FormulaC10H16O6
InChI KeyAGZPNUZBDCYTBB-UHFFFAOYSA-N
SMILESCCOC(=O)C(C(=O)OCC)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid after melting
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only