Triethyl orthoformate, 98%
Triethyl orthoformate, 98%
Triethyl orthoformate, 98%
Thermo Scientific Chemicals

Triethyl orthoformate, 98%

CAS: 122-51-0 | C7H16O3 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 139660025
Price (EUR)
130,00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
130,00
Each
Chemical Identifiers
CAS122-51-0
IUPAC Name(diethoxymethoxy)ethane
Molecular FormulaC7H16O3
InChI KeyGKASDNZWUGIAMG-UHFFFAOYSA-N
SMILESCCOC(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3900 to 1.3920 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only