Triethyl phosphite, 98%, Thermo Scientific Chemicals
Triethyl phosphite, 98%, Thermo Scientific Chemicals
Triethyl phosphite, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl phosphite, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 139691000
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS122-52-1
IUPAC Nametriethyl phosphite
Molecular FormulaC6H15O3P
InChI KeyBDZBKCUKTQZUTL-UHFFFAOYSA-N
SMILESCCOP(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only