Trifluoroacetic acid, 99+%, extra pure
Trifluoroacetic acid, 99+%, extra pure
Trifluoroacetic acid, 99+%, extra pure
Thermo Scientific Chemicals

Trifluoroacetic acid, 99+%, extra pure

CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
檢視全部
Glass Bottle
產品號碼 139720250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
化學識別
CAS76-05-1
IUPAC Nametrifluoroacetic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)F
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Water=<0.3 % (Coulometric)
Color scale=<20 APHA
Titration with NaOH99.0 to 101.0 %
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only