2',4',6'-Trihydroxyacetophenone, 98%
2',4',6'-Trihydroxyacetophenone, 98%
2',4',6'-Trihydroxyacetophenone, 98%
Thermo Scientific Chemicals

2',4',6'-Trihydroxyacetophenone, 98%

CAS: 480-66-0 | C8H8O4 | 168.15 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 139790250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS480-66-0
IUPAC Name1-(2,4,6-trihydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O4
InChI KeyXLEYFDVVXLMULC-UHFFFAOYSA-N
SMILESCC(=O)C1=C(O)C=C(O)C=C1O
さらに表示
Appearance (Color)Yellow to orange to tan
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point217°C to 222°C
HPLC>=97.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only