1,2,6-Hexanetriol, 97+%, extra pure
1,2,6-Hexanetriol, 97+%, extra pure
1,2,6-Hexanetriol, 97+%, extra pure
Thermo Scientific Chemicals

1,2,6-Hexanetriol, 97+%, extra pure

CAS: 106-69-4 | C6H14O3 | 134.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
139835000500 gGlass bottle
Catalog number 139835000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-69-4
IUPAC Namehexane-1,2,6-triol
Molecular FormulaC6H14O3
InChI KeyZWVMLYRJXORSEP-UHFFFAOYNA-N
SMILESOCCCCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
AppearanceMay solidify at about 32°C
Infrared spectrumConforms
GC>=97.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only