Trimethylacetamide, 98%
Trimethylacetamide, 98%
Trimethylacetamide, 98%
Thermo Scientific Chemicals

Trimethylacetamide, 98%

CAS: 754-10-9 | C5H11NO | 101.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 140010250
Price (USD)
92.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
92.00
Each
Chemical Identifiers
CAS754-10-9
IUPAC Name2,2-dimethylpropanamide
Molecular FormulaC5H11NO
InChI KeyXIPFMBOWZXULIA-UHFFFAOYSA-N
SMILESCC(C)(C)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point152°C to 156°C
Appearance (Color)White to light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only