Trimetilacetamida, 98 %, Thermo Scientific Chemicals
Trimetilacetamida, 98 %, Thermo Scientific Chemicals
Trimetilacetamida, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimetilacetamida, 98 %, Thermo Scientific Chemicals

CAS: 754-10-9 | C5H11NO | 101.15 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Frasco de plástico
Número de catálogo 140011000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS754-10-9
IUPAC Name2,2-dimethylpropanamide
Molecular FormulaC5H11NO
InChI KeyXIPFMBOWZXULIA-UHFFFAOYSA-N
SMILESCC(C)(C)C(N)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point152°C to 156°C
Appearance (Color)White to light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only