Trimethylacetyl chloride, 99%
Trimethylacetyl chloride, 99%
Trimethylacetyl chloride, 99%
Thermo Scientific Chemicals

Trimethylacetyl chloride, 99%

CAS: 3282-30-2 | C5H9ClO | 120.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 140050010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3282-30-2
IUPAC Name2,2-dimethylpropanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyJVSFQJZRHXAUGT-UHFFFAOYSA-N
SMILESCC(C)(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration RCOCl98.5 to 101.5 %
Appearance (Color)Clear colorless to light pink
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4110 to 1.4130 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only