Trimetilhidroquinona, 97 %, Thermo Scientific Chemicals
Trimetilhidroquinona, 97 %, Thermo Scientific Chemicals
Trimetilhidroquinona, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimetilhidroquinona, 97 %, Thermo Scientific Chemicals

CAS: 700-13-0 | C9H12O2 | 152.19 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
Envase:
Frasco de plástico
Número de catálogo 140151000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS700-13-0
IUPAC Name2,3,5-trimethylbenzene-1,4-diol
Molecular FormulaC9H12O2
InChI KeyAUFZRCJENRSRLY-UHFFFAOYSA-N
SMILESCC1=CC(O)=C(C)C(C)=C1O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to beige to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point168°C to 175°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only