Trimethyl phosphonoacetate, 98%
Trimethyl phosphonoacetate, 98%
Trimethyl phosphonoacetate, 98%
Thermo Scientific Chemicals

Trimethyl phosphonoacetate, 98%

CAS: 5927-18-4 | C5H11O5P | 182.11 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 140235000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5927-18-4
IUPAC Namemethyl 2-(dimethoxyphosphoryl)acetate
Molecular FormulaC5H11O5P
InChI KeySIGOIUCRXKUEIG-UHFFFAOYSA-N
SMILESCOC(=O)CP(=O)(OC)OC
さらに表示
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.4360 to 1.4380 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only