HBSS, calcium, magnesium, no phenol red
Gibco™

HBSS, calcium, magnesium, no phenol red

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1402513410 x 500 mL
14025092500 mL
140250761000 mL
140251266 x 1000 mL
Catalog number 14025134
Price (USD)
279.65
Online exclusive
298.00
Save 18.35
Each
Quantity:
10 x 500 mL
Customize this product
Price (USD)
279.65
Online exclusive
298.00
Save 18.35
Each

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media, or for reagent preparation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Calcium, Magnesium, Glucose

Without: Phenol Red, Sodium Pyruvate

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialHanks' Balanced Salt Solution (HBSS)
ColorClear
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 to 305 mOsm/kg
Recommended StorageStorage conditions: 15°C to 30°C, Shipping conditions: Room temperature, Shelf life: 36 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration1X
For Use With (Application)Mammalian Cell Culture
GradeCell Culture
Physical FormLiquid
Product LineGibco
Quantity10 x 500 mL
Solution TypepH Buffer, Storage Solution and Cleaning Solution
pH6.7 to 7.8
Unit SizeEach

Frequently asked questions (FAQs)

What is the pH of HBSS, calcium, magnesium, no phenol red at room temperature.

At room temperature, HBSS, calcium, magnesium, no phenol red has a pH of 6.7 - 7.8.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?

The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why do you offer HBSS with and without phenol red?

Phenol red is a pH indicator which allows you to determine the optimal atmospheric conditions for your cell line during your particular assays. We offer both versions to allow you the option to utilize this indicator in your media.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can I use HBSS (with calcium and magnesium) to wash my cells before dissociation?

No, a saline solution that contains calcium and magnesium should not be used to wash cells before dissociation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is a precipitate in HBSS (with calcium and magnesium) normal?

No, this is not normal and if there is a precipitate in the product, it cannot be used. If the precipitate will not go back into solution, please contact Technical Support at techsupport@thermofisher.com for a replacement.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Production, concentration and titration of pseudotyped HIV-1-based lentiviral vectors.
Authors:Kutner RH, Zhang XY, Reiser J,
Journal:Nat Protoc
PubMed ID:19300443
'Over the past decade, lentiviral vectors have emerged as powerful tools for transgene delivery. The use of lentiviral vectors has become commonplace and applications in the fields of neuroscience, hematology, developmental biology, stem cell biology and transgenesis are rapidly emerging. Also, lentiviral vectors are at present being explored in the ... More