Triphenylamine, 99+%
Triphenylamine, 99+%
Triphenylamine, 99+%
Thermo Scientific Chemicals

Triphenylamine, 99+%

CAS: 603-34-9 | C18H15N | 245.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 140311000
Price (EUR)
181,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
181,00
Each
Chemical Identifiers
CAS603-34-9
IUPAC NameN,N-diphenylaniline
Molecular FormulaC18H15N
InChI KeyODHXBMXNKOYIBV-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Melting point125°C to 127°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only