Triphenylene, 98%
Triphenylene, 98%
Triphenylene, 98%
Thermo Scientific Chemicals

Triphenylene, 98%

CAS: 217-59-4 | C18H12 | 228.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 140360050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS217-59-4
IUPAC Nametriphenylene
Molecular FormulaC18H12
InChI KeySLGBZMMZGDRARJ-UHFFFAOYSA-N
SMILESC1=CC=C2C(=C1)C1=CC=CC=C1C1=CC=CC=C21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or needles
Melting point196°C to 200°C
Appearance (Color)White to yellow to beige
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only