Search
Search
| Catalog Number | Quantity | Osmolality |
|---|---|---|
| 14040091 also known as 14040-091 | 500 mL | 270 to 310 mOsm/kg |
| 14040083 also known as 14040-083 | 100 mL | 270 to 310 mOsm/kg |
| 14040141 | 100 mL | 270 to 300 mOsm/kg |
| 14040166 also known as 14040-166 | 20 x 100 mL | 270 to 310 mOsm/kg |
| 14040133 | 500 mL | 270 to 300 mOsm/kg |
| 14040174 also known as 14040-174 | 10 x 500 mL | 270 to 310 mOsm/kg |
| 14040117 | 1000 mL | 270 to 300 mOsm/kg |
| 14040224 also known as 14040-224 | 6 x 1000 mL | 270 to 310 mOsm/kg |
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red