Triphenylmethyl chloride, 98%
Triphenylmethyl chloride, 98%
Triphenylmethyl chloride, 98%
Thermo Scientific Chemicals

Triphenylmethyl chloride, 98%

CAS: 76-83-5 | C19H15Cl | 278.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 140401000
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.60
Save 7.95
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.60
Save 7.95
Each
Chemical Identifiers
CAS76-83-5
IUPAC Name(chlorodiphenylmethyl)benzene
Molecular FormulaC19H15Cl
InChI KeyJBWKIWSBJXDJDT-UHFFFAOYSA-N
SMILESClC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to yellow to light beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point109°C to 113°C
Titration Argentometric>=97.5 % (ex Cl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only