Triphenylphosphine oxide, 99%
Triphenylphosphine oxide, 99%
Triphenylphosphine oxide, 99%
Thermo Scientific Chemicals

Triphenylphosphine oxide, 99%

CAS: 791-28-6 | C18H15OP | 278.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 140431000
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.60
Save 11.95
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.60
Save 11.95
Each
Chemical Identifiers
CAS791-28-6
IUPAC Name(diphenylphosphoroso)benzene
Molecular FormulaC18H15OP
InChI KeyFIQMHBFVRAXMOP-UHFFFAOYSA-N
SMILESO=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink-brown
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point154°C to 158°C
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only