Triphenyl phosphite, 99%
Triphenyl phosphite, 99%
Triphenyl phosphite, 99%
Thermo Scientific Chemicals

Triphenyl phosphite, 99%

CAS: 101-02-0 | C18H15O3P | 310.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Plastic drum
Catalog number 140440100
Price (USD)
450.65
Online Exclusive
500.00
Save 49.35 (10%)
Each
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
450.65
Online Exclusive
500.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS101-02-0
IUPAC Nametriphenyl phosphite
Molecular FormulaC18H15O3P
InChI KeyHVLLSGMXQDNUAL-UHFFFAOYSA-N
SMILESO(P(OC1=CC=CC=C1)OC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Acid value=<0.5 mg KOH/g
Appearance (Color)Colorless to white to light yellow
Infrared spectrumConforms
Refractive index1.5890 to 1.5920 (20°C, 589 nm)
Appearance (Form)Clear liquid or low melting solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only