This product ships via ground from US northeast.
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99+%, for biochemistry

Tris(hydroxymethyl)aminomethane, 99+%, C4H11NO3, CAS Number-77-86-1, 2-amino-2-hydroxymethyl propane-1,3-diol, tham, tris, trisamine, tris base, tris buffer, tris hydroxymethyl aminomethane, trizma, trometamol, tromethamine, 2.5kg | CAS: 77-86-1 | C4H11NO3 | 121.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 140500100
Price (USD)
729.65
Online exclusive
810.00
Save 80.35
Each
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
729.65
Online exclusive
810.00
Save 80.35
Each
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 290 nm A: =<0.2
UV(40 % in water) 1 cm cell
Color of solution=<25 APHA (20 % in water)
Titration with HClO4>=99.0 %
Heavy metals (as Pb)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only