Tropolone, 98%
Tropolone, 98%
Tropolone, 98%
Thermo Scientific Chemicals

Tropolone, 98%

CAS: 533-75-5 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 140580010
Price (EUR)
43,00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,00
Each
Chemical Identifiers
CAS533-75-5
IUPAC Name2-hydroxycyclohepta-2,4,6-trien-1-one
Molecular FormulaC7H6O2
InChI KeyMDYOLVRUBBJPFM-UHFFFAOYSA-N
SMILESOC1=CC=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
GC>=97.5 %
Appearance (Form)Crystalline powder
Melting point50°C to 54°C (on dry substance)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only