Tiramina, 97 %, Thermo Scientific Chemicals
Tiramina, 97 %, Thermo Scientific Chemicals
Tiramina, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tiramina, 97 %, Thermo Scientific Chemicals

CAS: 51-67-2 | C8H11NO | 137.18 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 140610250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS51-67-2
IUPAC Name4-(2-aminoethyl)phenol
Molecular FormulaC8H11NO
InChI KeyDZGWFCGJZKJUFP-UHFFFAOYSA-N
SMILESNCCC1=CC=C(O)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Off-white to beige to light brown
Infrared spectrumConforms
Melting point158°C to 163°C
Appearance (Form)Powder
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only