2-Undecanone, 98%
2-Undecanone, 98%
2-Undecanone, 98%
Thermo Scientific Chemicals

2-Undecanone, 98%

CAS: 112-12-9 | C11H22O | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 140692500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-12-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4280 to 1.4310 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only