Uridine, 99%
Uridine, 99%
Uridine, 99%
Thermo Scientific Chemicals

Uridine, 99%

CAS: 58-96-8 | C9H12N2O6 | 244.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 140771000
Price (USD)
320.65
Special offer
Online exclusive
Ends: 28-Dec-2026
377.00
Save 56.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
320.65
Special offer
Online exclusive
Ends: 28-Dec-2026
377.00
Save 56.35
Each
Chemical Identifiers
CAS58-96-8
IUPAC Name1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC9H12N2O6
InChI KeyDRTQHJPVMGBUCF-UHFFFAOYNA-N
SMILESOCC1OC(C(O)C1O)N1C=CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrophotometry>=98 % (lambda max. 262 nm, pH 7)
Loss on drying=<1 % (105°C, 2 hrs)
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only