Vinyl acetate, 99+%, stabilized
Vinyl acetate, 99+%, stabilized
Vinyl acetate, 99+%, stabilized
Thermo Scientific Chemicals

Vinyl acetate, 99+%, stabilized

CAS: 108-05-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 140840025
Price (USD)
71.65
Online exclusive
79.70
Save 8.05
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
71.65
Online exclusive
79.70
Save 8.05
Each
Chemical Identifiers
CAS108-05-4
IUPAC Nameethenyl acetate
Molecular FormulaC4H6O2
InChI KeyXTXRWKRVRITETP-UHFFFAOYSA-N
SMILESCC(=O)OC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Acidity (CH3COOH)=<0.005 %
Refractive index1.3940 to 1.3960 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only