Vinylene carbonate, 98%, stabilized
Vinylene carbonate, 98%, stabilized
Vinylene carbonate, 98%, stabilized
Vinylene carbonate, 98%, stabilized
Thermo Scientific Chemicals

Vinylene carbonate, 98%, stabilized

CAS: 872-36-6 | C3H2O3 | 86.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 140890050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS872-36-6
IUPAC Name2H-1,3-dioxol-2-one
Molecular FormulaC3H2O3
InChI KeyVAYTZRYEBVHVLE-UHFFFAOYSA-N
SMILESO=C1OC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Stabilizer=<0.1 % BHT
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4200 to 1.4230 (20°C, 589 nm)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only